Anti-IGF1 Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB134140-100EA 835 EUR
ABCAAB134140-100
Anti-IGF1 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IGF1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen