Anti-LRP6 Rabbit Monoclonal Antibody [clone: EPR2423]
Lieferant: Abcam
ABCAAB134146-100EA 934 EUR
ABCAAB134146-100
Anti-LRP6 Rabbit Monoclonal Antibody [clone: EPR2423]
Antikörper
Type: Primary
Antigen: LRP6 (low density lipoprotein receptor-related protein 6)
Clonality: monoclonal
Clone: EPR2423
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen