Anti-SIRT2 Rabbit Monoclonal Antibody [clone: EPR1667]

Lieferant: Abcam
ABCAAB134171-100EA 939 EUR
ABCAAB134171-100
Anti-SIRT2 Rabbit Monoclonal Antibody [clone: EPR1667]
Antikörper
Type: Primary
Antigen: SIRT2 (sirtuin 2)
Clonality: monoclonal
Clone: EPR1667
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR