Anti-CCND1 Rabbit monoclonal antibody unconjugated [clone: EPR2241]

Lieferant: Abcam
ABCAAB134175-100EA 950 EUR
ABCAAB134175-100
Anti-CCND1 Rabbit monoclonal antibody unconjugated [clone: EPR2241]
Antikörper
Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: monoclonal
Clone: EPR2241
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR