Anti-ERBB2 Rabbit Monoclonal Antibody [clone: EP1045Y]

Lieferant: Abcam
ABCAAB134182-100EA 935 EUR
ABCAAB134182-100
Anti-ERBB2 Rabbit Monoclonal Antibody [clone: EP1045Y]
Antikörper
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: monoclonal
Clone: EP1045Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR