Anti-KDR Rabbit Monoclonal Antibody [clone: EPRER16Y]

Lieferant: Abcam
ABCAAB134191-100EA 854 EUR
ABCAAB134191-100
Anti-KDR Rabbit Monoclonal Antibody [clone: EPRER16Y]
Antikörper
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: monoclonal
Clone: EPRER16Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR