Anti-CD63 Mouse Monoclonal Antibody (Biotin) [clone: MEM-259]

Lieferant: Abcam
ABCAAB134331-100EA 834 EUR
ABCAAB134331-100
Anti-CD63 Mouse Monoclonal Antibody (Biotin) [clone: MEM-259]
Antikörper
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone: MEM-259
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR