Anti-CD86 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BU63]

Lieferant: Abcam
AB134385-50
ABCAAB134385-50EA 876 EUR
ABCAAB134385-50
Anti-CD86 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BU63]
Antikörper
Type: Primary
Antigen: CD86 (CD86 molecule)
Clonality: monoclonal
Clone: BU63
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR