Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]

Lieferant: Abcam
ABCAAB134399-50EA 407 EUR
ABCAAB134399-50
Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]
Antikörper
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: monoclonal
Clone: HIT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR