Anti-CD40LG Mouse Monoclonal Antibody (Biotin) [clone: 24-31]

Lieferant: Abcam
AB134407-50
ABCAAB134407-50EA 876 EUR
ABCAAB134407-50
Anti-CD40LG Mouse Monoclonal Antibody (Biotin) [clone: 24-31]
Antikörper
Type: Primary
Antigen: CD40LG (CD40 ligand)
Clonality: monoclonal
Clone: 24-31
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR