Anti-GFAP Chicken Polyclonal Antibody

Lieferant: Abcam
AB134436-100
ABCAAB134436-100EA 891 EUR
ABCAAB134436-100
Anti-GFAP Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR