Anti-CD28 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: CD28.2]
Lieferant: Abcam
ABCAAB134472-50EA 755 EUR
ABCAAB134472-50
Anti-CD28 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: CD28.2]
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone: CD28.2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen