Anti-UPF3B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB134566-100EA 835 EUR
ABCAAB134566-100
Anti-UPF3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UPF3B (UPF3 regulator of nonsense transcripts homolog B (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR