Anti-MIP Rabbit Polyclonal Antibody

Lieferant: Abcam
AB134695-100
ABCAAB134695-100EA 882 EUR
ABCAAB134695-100
Anti-MIP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MIP (major intrinsic protein of lens fiber)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR