Anti-REN Mouse Monoclonal Antibody [clone: 7D3-E3]

Lieferant: Abcam
ABCAAB134783-50EA 840 EUR
ABCAAB134783-50
Anti-REN Mouse Monoclonal Antibody [clone: 7D3-E3]
Antikörper
Type: Primary
Antigen: REN (renin)
Clonality: monoclonal
Clone: 7D3-E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR