Anti-INADL Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB134784-50EA 882 EUR
ABCAAB134784-50
Anti-INADL Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: INADL (InaD-like (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR