Anti-PITX3 Rabbit Monoclonal Antibody

Lieferant: Abcam
AB134920-100
ABCAAB134920-100EA 777 EUR
ABCAAB134920-100
Anti-PITX3 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PITX3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR