Anti-CHMP4B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB135154-100EA 835 EUR
ABCAAB135154-100
Anti-CHMP4B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHMP4B (charged multivesicular body protein 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR