Anti-PPM1A Mouse Monoclonal Antibody [clone: 7F12]

Lieferant: Abcam
AB135249-100
ABCAAB135249-100EA 994 EUR
ABCAAB135249-100
Anti-PPM1A Mouse Monoclonal Antibody [clone: 7F12]
Antikörper
Type: Primary
Antigen: PPM1A (protein phosphatase, Mg2+/Mn2+ dependent, 1A)
Clonality: monoclonal
Clone: 7F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR