Anti-PI3K Rabbit Monoclonal Antibody

Lieferant: Abcam
ABCAAB135371-7EA 814 EUR
ABCAAB135371-7
Anti-PI3K Rabbit Monoclonal Antibody
Type: Primary
Antigen: PI3K
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR