Anti-ATF6 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB135707-400EA 780 EUR
ABCAAB135707-400
Anti-ATF6 Rabbit Polyclonal Antibody
Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR