Anti-SEMA4C Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB135856-100EA 797 EUR
ABCAAB135856-100
Anti-SEMA4C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEMA4C (sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR