Anti-CAMKK2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB135979-100EA 816 EUR
ABCAAB135979-100
Anti-CAMKK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMKK2 (calcium/calmodulin-dependent protein kinase kinase 2, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR