Anti-CLCNKA Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB136039-100EA 797 EUR
ABCAAB136039-100
Anti-CLCNKA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLCNKA (chloride channel, voltage-sensitive Ka)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR