Anti-ARHGEF3 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB136064-100EA 797 EUR
ABCAAB136064-100
Anti-ARHGEF3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARHGEF3 (Rho guanine nucleotide exchange factor (GEF) 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human