Anti-GPR174 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB136271-100EA 840 EUR
ABCAAB136271-100
Anti-GPR174 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPR174 (G protein-coupled receptor 174)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human