Anti-S100 Rabbit Monoclonal Antibody [clone: D28-E]
Lieferant: Abcam
ABCAAB136629-250EA 852 EUR
ABCAAB136629-250
Anti-S100 Rabbit Monoclonal Antibody [clone: D28-E]
Antikörper
Type: Primary
Antigen: S100
Clonality: monoclonal
Clone: D28-E
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human