Anti-RHBG Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB136658-50EA 797 EUR
ABCAAB136658-50
Anti-RHBG Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RHBG (Rh family, B glycoprotein (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR