Anti-DAG1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB136665-50EA 835 EUR
ABCAAB136665-50
Anti-DAG1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DAG1 (dystroglycan 1 (dystrophin-associated glycoprotein 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR