Anti-HLA-DQA2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB136667-50EA 797 EUR
ABCAAB136667-50
Anti-HLA-DQA2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HLA-DQA2 (major histocompatibility complex, class II, DQ alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR