Anti-IgG Rabbit Polyclonal Antibody (bGal (beta Galactosidase))

Lieferant: Abcam
AB136710-500
ABCAAB136710-500EA 445 EUR
ABCAAB136710-500
Anti-IgG Rabbit Polyclonal Antibody (bGal (beta Galactosidase))
Antikörper
Type: Secondary
Antigen: IgG (Bovine) (bovine IgG)
Clonality: polyclonal
Clone:
Conjugation: beta-Galactosidase
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR