Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB136742-500EA 448 EUR
ABCAAB136742-500
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (Dog) (dog IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR