Anti-IgG Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB136748-1EA 445 EUR
ABCAAB136748-1
Anti-IgG Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR