Anti-HSD3B7 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB136846-100EA 797 EUR
ABCAAB136846-100
Anti-HSD3B7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSD3B7 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR