Anti-PLXNB1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB136880-50
ABCAAB136880-50EA 882 EUR
ABCAAB136880-50
Anti-PLXNB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLXNB1 (plexin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR