Anti-ACTB Rabbit Monoclonal Antibody [clone: S12-I]
Lieferant: Abcam
ABCAAB136905-100EA 840 EUR
ABCAAB136905-100
Anti-ACTB Rabbit Monoclonal Antibody [clone: S12-I]
Antikörper
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone: S12-I
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse