Anti-CD1A Rabbit Monoclonal Antibody [clone: L21-A]

Lieferant: Abcam
ABCAAB136922-250EA 859 EUR
ABCAAB136922-250
Anti-CD1A Rabbit Monoclonal Antibody [clone: L21-A]
Antikörper
Type: Primary
Antigen: CD1A (CD1a molecule)
Clonality: monoclonal
Clone: L21-A
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR