Anti-IL17RC Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB136991-100EA 840 EUR
ABCAAB136991-100
Anti-IL17RC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL17RC (interleukin 17 receptor C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR