Anti-RPL26L1 Rabbit Monoclonal Antibody

Lieferant: Abcam
ABCAAB137046-100EA 856 EUR
ABCAAB137046-100
Anti-RPL26L1 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RPL26L1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR