Anti-Mitofilin Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB137057-100EA 866 EUR
ABCAAB137057-100
Anti-Mitofilin Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Mitofilin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen