Anti-SOS2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137199-100EA 797 EUR
ABCAAB137199-100
Anti-SOS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SOS2 (son of sevenless homolog 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR