Anti-MAP3K5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137339-50EA 840 EUR
ABCAAB137339-50
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP3K5 (mitogen-activated protein kinase kinase kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR