Anti-HDAC7 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB137366-50EA 859 EUR
ABCAAB137366-50
Anti-HDAC7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HDAC7 (histone deacetylase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human