Anti-RPSA Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137388-100EA 859 EUR
ABCAAB137388-100
Anti-RPSA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPSA (ribosomal protein SA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR