Anti-CDYL Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB137423-100EA 840 EUR
ABCAAB137423-100
Anti-CDYL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDYL (chromodomain protein, Y-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human