Anti-Glutathione Peroxidase 2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137431-100EA 859 EUR
ABCAAB137431-100
Anti-Glutathione Peroxidase 2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Glutathione Peroxidase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR