Anti-MMP12 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137444-100EA 859 EUR
ABCAAB137444-100
Anti-MMP12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP12 (matrix metallopeptidase 12 (macrophage elastase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR