Anti-BTK Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137504-50EA 840 EUR
ABCAAB137504-50
Anti-BTK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BTK (Bruton agammaglobulinemia tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR