Anti-PAG3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137514-100EA 797 EUR
ABCAAB137514-100
Anti-PAG3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAG3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR