Anti-CXCL14 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137541-50
ABCAAB137541-50EA 895 EUR
ABCAAB137541-50
Anti-CXCL14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CXCL14 (chemokine (C-X-C motif) ligand 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR